ContactUsLinkComponent
For quantitative detection of human Persephin in cell culture supernates, serum and plasma(heparin, EDTA). 1 * 1 KIT
Catalog # BSBTEK1509
Supplier:  Boster Bio
For quantitative detection of human Persephin in cell culture supernates, serum and plasma(heparin, EDTA). 1 * 1 KIT
Catalog # BSBTEK1509
Supplier:  Boster Bio