ContactUsLinkComponent
For quantitative detection of mouse FCRN in cell culture supernates, serum, plasma(heparin, EDTA) and cell lysates. 1 * 1 KIT
Catalog # BSBTEK1454
Supplier:  Boster Bio
For quantitative detection of mouse FCRN in cell culture supernates, serum, plasma(heparin, EDTA) and cell lysates. 1 * 1 KIT
Catalog # BSBTEK1454
Supplier:  Boster Bio