ContactUsLinkComponent
Minichiller 600 Temperature range: -20 ... 40 C Cooling capacity (15C): 0,6 kW Cooling capacity (0C): 0,5 kW Pressure pump max.: 33 l/min / 0.7 bar Suction pump max. : 22 l/min / 0,4 bar Pump connection: M16x1 male Power supply: 115V 1 60Hz With 1 * 1 items
  HUBE3006.0073.99
 :  HUBER
Minichiller 600 Temperature range: -20 ... 40 C Cooling capacity (15C): 0,6 kW Cooling capacity (0C): 0,5 kW Pressure pump max.: 33 l/min / 0.7 bar Suction pump max. : 22 l/min / 0,4 bar Pump connection: M16x1 male Power supply: 115V 1 60Hz With 1 * 1 items
  HUBE3006.0073.99
 :  HUBER
Frequently Bought Together