ContactUsLinkComponent
CREB1 - Human, 4 unique 29mer shRNA constructs in retroviral GFP vector (Gene ID = 1385). 5µg purified plasmid DNA per construct Non-effective 29-mer scrambled shRNA cassette in pGFP-V-RS Vector, TR30013, included for free. 1 * 1 KIT
Catalog # ORIGTG313740
Supplier:  OriGene
CREB1 - Human, 4 unique 29mer shRNA constructs in retroviral GFP vector (Gene ID = 1385). 5µg purified plasmid DNA per construct Non-effective 29-mer scrambled shRNA cassette in pGFP-V-RS Vector, TR30013, included for free. 1 * 1 KIT
Catalog # ORIGTG313740
Supplier:  OriGene
Frequently Bought Together