ContactUsLinkComponent
Burette with straight stopcock, PTFE key, with Schellbach stripe, class USP 25 ml (0,1), logo VOLAC-FORTUNA, amber grad. With Certi ficate 1 * 1 items
Catalog # POULUS9806/WAC/G
Supplier:  Poulten & Graf
Burette with straight stopcock, PTFE key, with Schellbach stripe, class USP 25 ml (0,1), logo VOLAC-FORTUNA, amber grad. With Certi ficate 1 * 1 items
Catalog # POULUS9806/WAC/G
Supplier:  Poulten & Graf
Frequently Bought Together