ContactUsLinkComponent
Jalas Boot Safety 1878 Offroad, Wide fit with Aluminium Toecap & Steel Midsole, PU-Coated Leather Upper, Fleece Liner, PU Pronose, N itrile Rubber Outsole, Oil-resistant outsole, water repellant, anti-static properties, ESD, double shock absorp zones 1 * 1 PAIR
Catalog # EJEN1878-49
Supplier:  EJENDALS
undefined
Jalas Boot Safety 1878 Offroad, Wide fit with Aluminium Toecap & Steel Midsole, PU-Coated Leather Upper, Fleece Liner, PU Pronose, N itrile Rubber Outsole, Oil-resistant outsole, water repellant, anti-static properties, ESD, double shock absorp zones 1 * 1 PAIR
Catalog # EJEN1878-49
Supplier:  EJENDALS