ContactUsLinkComponent
pH sensor with pre-pressurized electrolyte, Cleaning: SIP, CIP, Autoclavation, no cable, VP 6 connector head, Diaphragm: HP Coatrami c, Material: glass shaft, Electrolyte: Phermlyte, Length: 225 mm, Diameter: 12 mm, Process temperature: 0 to 140 °C, 1 * 1 items
Catalog # HAMI238633-1642
Supplier:  HAMILTON BONADUZ
pH sensor with pre-pressurized electrolyte, Cleaning: SIP, CIP, Autoclavation, no cable, VP 6 connector head, Diaphragm: HP Coatrami c, Material: glass shaft, Electrolyte: Phermlyte, Length: 225 mm, Diameter: 12 mm, Process temperature: 0 to 140 °C, 1 * 1 items
Catalog # HAMI238633-1642
Supplier:  HAMILTON BONADUZ
Frequently Bought Together