ContactUsLinkComponent
Programmable digital Muffle Furnace, type FHP-12, with 4-side heating & programmable digital PID control system, temp. range: 300C - 1200C, capacity: 12 Liter, internal: 200 x 300 x 200 mm, ext.: 480 x 550 x 660 mm, power: 3,6 kW, power supply: 400 1 * 1 items
Catalog # WITGDH.WFH23.P12
Supplier:  witeg Labortechnik
Programmable digital Muffle Furnace, type FHP-12, with 4-side heating & programmable digital PID control system, temp. range: 300C - 1200C, capacity: 12 Liter, internal: 200 x 300 x 200 mm, ext.: 480 x 550 x 660 mm, power: 3,6 kW, power supply: 400 1 * 1 items
Catalog # WITGDH.WFH23.P12
Supplier:  witeg Labortechnik
Frequently Bought Together