ContactUsLinkComponent
Water bath shaking 45l, temp range: +5C to 100C, reciprocating motion, delay & continuous function incl., incl. stainless steel sp ring rack, 230V 50/60Hz Hz, with digital fuzzy control, type WSB-45 1 * 1 items
Catalog # WITGDH.WSB03045
Supplier:  witeg Labortechnik
Water bath shaking 45l, temp range: +5C to 100C, reciprocating motion, delay & continuous function incl., incl. stainless steel sp ring rack, 230V 50/60Hz Hz, with digital fuzzy control, type WSB-45 1 * 1 items
Catalog # WITGDH.WSB03045
Supplier:  witeg Labortechnik
Frequently Bought Together