ContactUsLinkComponent
Cryo Boxes Ratiolab® with Slip-Lid, Format 133 x 133 mm, polypropylene, natural color. Adjustable in height from 50 mm to 75 mm, autoclavable, grid 7 x 7. For tube Ø 17 mm. 1 * 5 items
Catalog # RATI5320310
Supplier:  RATIOLAB
Cryo Boxes Ratiolab® with Slip-Lid, Format 133 x 133 mm, polypropylene, natural color. Adjustable in height from 50 mm to 75 mm, autoclavable, grid 7 x 7. For tube Ø 17 mm. 1 * 5 items
Catalog # RATI5320310
Supplier:  RATIOLAB
Frequently Bought Together