ContactUsLinkComponent
This product is prepared by labeling highly cross-adsorbed goat anti-mouse IgG (H+L) with the green fluorescent dye CF™532. To mini mize cross-reactivity, the antibody has been adsorbed against human, bovine, horse, rabbit, and swine serum. 1 * 500 µl
Catalog # 20468-500UL
Supplier:  Biotium
This product is prepared by labeling highly cross-adsorbed goat anti-mouse IgG (H+L) with the green fluorescent dye CF™532. To mini mize cross-reactivity, the antibody has been adsorbed against human, bovine, horse, rabbit, and swine serum. 1 * 500 µl
Catalog # 20468-500UL
Supplier:  Biotium
Frequently Bought Together