ContactUsLinkComponent
This product is prepared by single labeling (DOL=1) highly cross-adsorbed goat anti-rabbit IgG (H+L) with CF™568ST for (d)-STORM sup er-resolution microscopy. To minimize cross-reactivity, the antibody has been adsorbed against human, mouse, and rat 1 * 50 µl
Catalog # 20801-50UL
Supplier:  Biotium
This product is prepared by single labeling (DOL=1) highly cross-adsorbed goat anti-rabbit IgG (H+L) with CF™568ST for (d)-STORM sup er-resolution microscopy. To minimize cross-reactivity, the antibody has been adsorbed against human, mouse, and rat 1 * 50 µl
Catalog # 20801-50UL
Supplier:  Biotium
Frequently Bought Together