ContactUsLinkComponent
- Diameter funnel approx. 120 mm - Maximum collection volume approx. 1.7 l - Hinged lid reduces escaping vapours to a minimum. - Lid lockable in open position - Dirt strainer can be removed without tools and can be disassembled for cleaning - Connect 1 * 1 items
Catalog # DUEH5.08.020
Supplier:  DUPERTHAL
- Diameter funnel approx. 120 mm - Maximum collection volume approx. 1.7 l - Hinged lid reduces escaping vapours to a minimum. - Lid lockable in open position - Dirt strainer can be removed without tools and can be disassembled for cleaning - Connect 1 * 1 items
Catalog # DUEH5.08.020
Supplier:  DUPERTHAL
Frequently Bought Together