- Diameter funnel approx. 120 mm - Hinged lid reduces escaping vapours to a minimum. - Lid lockable in open position - Dirt strainer can be removed without tools and can be disassembled for cleaning - Connection thread for adaption to bench penetrati 1 * 1 items
DUEH5.08.012
Frequently Bought Together