Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
GFP ELISA Kit

GFP ELISA Kit

GFP ELISA Kit, 96 tests
Katalogové číslo ABCAAB171581-96
Dodavatel:   Abcam
GFP ELISA Kit
Katalogové číslo ABCAAB171581-96
Dodavatel:  Abcam
Číslo CAS:  
GFP ELISA Kit
GFP ELISA Kit

GFP ELISA Kit, 96 tests

Přejít na:
Číslo CAS:  
Price
20 090,00 Kč
Přihlásit se pro dostupnost
1 * 96 Tests
Product Details & Documents

GFP ELISA kit is a single-wash 90-min SimpleStep ELISA® used to quantify Green Fluorescence protein with a sensitivity of 2.8 pg/ml. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. Colorimetric assay - 450 nm readout; works on any plate reader. Different formats for different needs: 10×96-well plates for bulk orders and 384-well for higher throughput. Broad sample compatibility, works with tissue lysate and cell lysate.

Documents

Specifikace

Specifikace

  • Description:
    GFP ELISA Kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Species independent
  • Sample type:
    Tissue Lysate, Cell Lysate
  • Detection range:
    18,8 - 1200 pg/ml
  • Storage temperature:
    4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 hr 30 min
  • Cat. No.:
    ABCAAB171581-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    2,8 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    GFP
  • Regulatory status:
    RUO

Specifikace

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.