RECOMBINANT MOUSE KDM4B / JMJD2B PROTEIN 1 * 20 µG
Katalogové číslo ABCAAB139634-20
Dodavatel: Abcam
RECOMBINANT MOUSE KDM4B / JMJD2B PROTEIN 1 * 20 µG
Katalogové číslo ABCAAB139634-20
Dodavatel: Abcam
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Sealing film for manual application, Parafilm® M
Dodavatel: HEATHROW
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
VWR® Facial Tissues
Dodavatel: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Dodavatel: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Dodavatel: VWR Collection
VWR® Serological Pipette, Standard Line
Dodavatel: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Dodavatel: KIMBERLY CLARK