Collapsible carts (steel), handlebar folds onto platform, platform size 900 x 600 mm, plate made of beech multiplex wood, 2 swivel castor / 2 fixed castor solid rubber wheel Ø 160 mm, capacity 300 kg, powder coated black-silver 1 * 1 items
Catalog # GORRK2P-723/03
Supplier: Gorr Transporttechnik GmbH
Frequently Bought Together