ContactUsLinkComponent
Steel-Bottle-Trolley for 1 steel bottle 10-40 litres, bottle support with chain, 2 pneumatic tyres 260 mm, heigt 1150 mm, powder coa ted black-silver 1 * 1 items
Catalog # GORRKKR-820/02
Steel-Bottle-Trolley for 1 steel bottle 10-40 litres, bottle support with chain, 2 pneumatic tyres 260 mm, heigt 1150 mm, powder coa ted black-silver 1 * 1 items
Catalog # GORRKKR-820/02
Frequently Bought Together