ContactUsLinkComponent
Euro-norm plastic bin 90 litres, euro-standard 800 x 600 x 220 mm, with closed bottom and closed side-walls 1 * 1 items
Catalog # GORRKBE-184/00
Euro-norm plastic bin 90 litres, euro-standard 800 x 600 x 220 mm, with closed bottom and closed side-walls 1 * 1 items
Catalog # GORRKBE-184/00
Frequently Bought Together