ContactUsLinkComponent
Conductivity cell with fixed cable, 4 pole cell, plastic shaft, 1.5 m cable with 8 pole plug, sensor material graphite, cell constan t 0.33 cm-1, temp.-sensor NTC 30 kOhm, length 120 mm, 12 mm Ø, -5...+80 °C 1 * 1 items
Catalog # SCOI285206420
Supplier:  SI Analytics
Conductivity cell with fixed cable, 4 pole cell, plastic shaft, 1.5 m cable with 8 pole plug, sensor material graphite, cell constan t 0.33 cm-1, temp.-sensor NTC 30 kOhm, length 120 mm, 12 mm Ø, -5...+80 °C 1 * 1 items
Catalog # SCOI285206420
Supplier:  SI Analytics
Frequently Bought Together