28 mm PP Closure for 30 mL bottle. Process: Particulate Cleaning & Autoclave. Packaging: Single autoclave bag each cap. 72 per case. Certification: C of A Endotoxin (Report Actual Results), C of A USP Particles, C of A USP Sterility, Certificate of P 1 * 72 items
Catalog # NALGCE-N6MM28PA-EP
Supplier: Thermo Fisher Scientific
Frequently Bought Together