ContactUsLinkComponent
CONDUCTIVITY CELL C = 0.7 for Metrohm conductivity WITH REF Integrated temperature sensor for analysis. 1 * 1 items
Catalog # METR6.0915.100
Supplier:  METROHM
CONDUCTIVITY CELL C = 0.7 for Metrohm conductivity WITH REF Integrated temperature sensor for analysis. 1 * 1 items
Catalog # METR6.0915.100
Supplier:  METROHM
Frequently Bought Together