ContactUsLinkComponent
XTip 20 µL Reload System Pipet Tips. Clean Pack Reloads, Pre-sterile, Rack Lid and Base, 96 Tip/Card, 10 Cards/Pack, 5 Pack/Case. P atented features: FlexFit™ for superior seal, Blade™ for accuracy, and lot traceable. 1 * 960 items
  613-5457
 :  BIOTIX
XTip 20 µL Reload System Pipet Tips. Clean Pack Reloads, Pre-sterile, Rack Lid and Base, 96 Tip/Card, 10 Cards/Pack, 5 Pack/Case. P atented features: FlexFit™ for superior seal, Blade™ for accuracy, and lot traceable. 1 * 960 items
  613-5457
 :  BIOTIX
Frequently Bought Together