ContactUsLinkComponent
XTip 20 µL Reload System Pipet Tips. Tip Eject Reload System, Pre-sterile, 96 Tip/Card, 10 Cards/Pack, 5 Pack/Case. Patented featur es: FlexFit™ for superior seal, Blade™ for accuracy, and lot traceable. 1 * 960 items
Catalog # 613-5453
Supplier:  BIOTIX
XTip 20 µL Reload System Pipet Tips. Tip Eject Reload System, Pre-sterile, 96 Tip/Card, 10 Cards/Pack, 5 Pack/Case. Patented featur es: FlexFit™ for superior seal, Blade™ for accuracy, and lot traceable. 1 * 960 items
Catalog # 613-5453
Supplier:  BIOTIX
Frequently Bought Together