XTip 20ul Pipet Tips for Rainin LTS pipettes. Racked, Pre-sterile, 10 racks of 96 Tips/Pack, 5 Packs/Case. Fits Rainin LTS pipette s ideal for both single & multichannel pipettors, graduation marks, Patented features: FlexFit for superior seal, Bla 1 * 960 items
Catalog # 613-5449
Supplier: BIOTIX
Frequently Bought Together