ContactUsLinkComponent
IntelliCal™ double Platinum probe with integral T-sensor, model MTC695, ±1200 mV, -10…80°C, 1 m cable with M12 plug for HQd meter series used for Karl Fischer titrations and other potentiometric and amperometric titrations with AT/KF1000 1 * 1 items
Catalog # HACHMTC69501
Supplier:  Hach
IntelliCal™ double Platinum probe with integral T-sensor, model MTC695, ±1200 mV, -10…80°C, 1 m cable with M12 plug for HQd meter series used for Karl Fischer titrations and other potentiometric and amperometric titrations with AT/KF1000 1 * 1 items
Catalog # HACHMTC69501
Supplier:  Hach
Frequently Bought Together