ContactUsLinkComponent
FLUORESCEIN - CONJUGATED RABBIT F(ab')2 FRAGMENT TO MOUSE IgG (WHOLE MOLECULE). Anti-Mouse Host: rabbit Form: lyophilized (F(ab)2 ) Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Con 1 * 2 mL
Catalog # ICNA0855505
Supplier:  MP Biomedicals
FLUORESCEIN - CONJUGATED RABBIT F(ab')2 FRAGMENT TO MOUSE IgG (WHOLE MOLECULE). Anti-Mouse Host: rabbit Form: lyophilized (F(ab)2 ) Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Con 1 * 2 mL
Catalog # ICNA0855505
Supplier:  MP Biomedicals
Frequently Bought Together