ContactUsLinkComponent
FLUORESCEIN - CONJUGATED GOAT F(ab')2 FRAGMENT TO HUMAN IgM (5Fc µ). Anti-Human Host: goat Form: lyophilized (F(ab´)2 ) Applications : Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugated 1 * 2 mL
Catalog # ICNA0855185
Supplier:  MP Biomedicals
FLUORESCEIN - CONJUGATED GOAT F(ab')2 FRAGMENT TO HUMAN IgM (5Fc µ). Anti-Human Host: goat Form: lyophilized (F(ab´)2 ) Applications : Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugated 1 * 2 mL
Catalog # ICNA0855185
Supplier:  MP Biomedicals
Frequently Bought Together