ContactUsLinkComponent
2,2 - AZINO - bis - (3 - ETHYLBENZTHIAZOLINE - 6 - SULFONIC ACID) SOLUTION. (ABTS) Stabilized Liquid Substrate A safe, convenient a nd good overall performing substrate for ELISA. It may be used for both kinetic and endpoint reactions. Supplied read 1 * 1 L
  ICNA04821810
2,2 - AZINO - bis - (3 - ETHYLBENZTHIAZOLINE - 6 - SULFONIC ACID) SOLUTION. (ABTS) Stabilized Liquid Substrate A safe, convenient a nd good overall performing substrate for ELISA. It may be used for both kinetic and endpoint reactions. Supplied read 1 * 1 L
  ICNA04821810
Frequently Bought Together