ContactUsLinkComponent
beta-AMYLOID, FRAGMENT 1-42. Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser -Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-AlaPurity: >98% 1 * 500 µG
Catalog # ICNA02159871.5
Supplier:  MP Biomedicals
beta-AMYLOID, FRAGMENT 1-42. Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser -Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-AlaPurity: >98% 1 * 500 µG
Catalog # ICNA02159871.5
Supplier:  MP Biomedicals
Frequently Bought Together