ContactUsLinkComponent
DOXORUBICIN HYDROCHLORIDE. Purity: >95% by HPLCChemotherapeutic agent. Antitumor, immunosuppressive, and antibiotic agent. Blocks RNA polymerase and reverse transcriptase. Also inhibits nucleic acid synthesis. 1 * 1 mg
Catalog # ICNA0215910101
Supplier:  MP Biomedicals
DOXORUBICIN HYDROCHLORIDE. Purity: >95% by HPLCChemotherapeutic agent. Antitumor, immunosuppressive, and antibiotic agent. Blocks RNA polymerase and reverse transcriptase. Also inhibits nucleic acid synthesis. 1 * 1 mg
Catalog # ICNA0215910101
Supplier:  MP Biomedicals
Frequently Bought Together