ContactUsLinkComponent
RIBONUCLEIC ACID TRANSFER. Activity: 19 A260 units/mg.Acceptor activities given in picomoles (10-12 moles) per A260 unit.Unit Defin ition: One unit will yield an A260 of 1.0 in 1.0 ml of water (1 cm light path).From Bakers YeastLyophilized powderTyp 1 * 1 KU
Catalog # ICNA0215653491
Supplier:  MP Biomedicals
RIBONUCLEIC ACID TRANSFER. Activity: 19 A260 units/mg.Acceptor activities given in picomoles (10-12 moles) per A260 unit.Unit Defin ition: One unit will yield an A260 of 1.0 in 1.0 ml of water (1 cm light path).From Bakers YeastLyophilized powderTyp 1 * 1 KU
Catalog # ICNA0215653491
Supplier:  MP Biomedicals
Frequently Bought Together