ContactUsLinkComponent
GOAT AFFINITY PURIFIED F(ab')2 FRAGMENT TO HUMAN IgM (5Fc MU). Anti-Human Host: goat Form: affinity purified liquid F(ab ')2 Applica tions: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 1 mg
Catalog # ICNA0856960
Supplier:  MP Biomedicals
GOAT AFFINITY PURIFIED F(ab')2 FRAGMENT TO HUMAN IgM (5Fc MU). Anti-Human Host: goat Form: affinity purified liquid F(ab ')2 Applica tions: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 1 mg
Catalog # ICNA0856960
Supplier:  MP Biomedicals
Frequently Bought Together