ContactUsLinkComponent
GENETICIN® DISULFATE SALT. Cell Culture GradeDisulfate SaltPurity: ~98%Geneticin is an aminoglycoside related to Gentamicin and is t oxic to bacteria, yeast, higher plants, mammalian cells, protozoans and helminths. The resistance genes are located o 1 * 250 mg
Catalog # ICNA0215878283
Supplier:  MP Biomedicals
GENETICIN® DISULFATE SALT. Cell Culture GradeDisulfate SaltPurity: ~98%Geneticin is an aminoglycoside related to Gentamicin and is t oxic to bacteria, yeast, higher plants, mammalian cells, protozoans and helminths. The resistance genes are located o 1 * 250 mg
Catalog # ICNA0215878283
Supplier:  MP Biomedicals
Frequently Bought Together