Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
2-Fenoxyethanol

2-Fenoxyethanol

2725, 500 mL
Catalog # ICNA0215014890
Supplier:   MP Biomedicals
2-Fenoxyethanol
Catalog # ICNA0215014890
Supplier:  MP Biomedicals
CAS Number:  122-99-6
2-Fenoxyethanol
2-Fenoxyethanol

2725, 500 mL

CAS Number:  122-99-6
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents

Important notice: this product belongs to MP Biomedicals rare chemical library. Most rare chemical products are available for immediate delivery and sold "As Is". MP biomedicals does not collect analytical data for those products. All sales are final. However, analytical test may be performed upon custom request for bulk orders.

Caution: Research Use Only (RUO).

Synonyms:  ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
  • CAS Number:122-99-6
  • Formula:C₆H₅OCH₂CH₂OH
  • MDL Number:MFCD00002857
  • Molecular Weight:138.17
  • Melting Pt:57 ° F (lit.)
  •  :Density: 1,1094 g/cm³ (20 °C)
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Identity Test:
    Passes
  • FTIR:
    Conforms to Standard

Specifications

  • Bal.:
    500 mL
  • Sekvence:
    2725

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.