ContactUsLinkComponent
LEUCINE AMINOPEPTIDASE. Source: Porcine Kidney Microsomes Form: Freeze-dried powder Solubility: Soluble in distilled water or dilute buffer Activity: > 5 U/mg solid Protein: 50% (biuret) 1 * 25 AU
Catalog # ICNA0215388525
Supplier:  MP Biomedicals
LEUCINE AMINOPEPTIDASE. Source: Porcine Kidney Microsomes Form: Freeze-dried powder Solubility: Soluble in distilled water or dilute buffer Activity: > 5 U/mg solid Protein: 50% (biuret) 1 * 25 AU
Catalog # ICNA0215388525
Supplier:  MP Biomedicals
Frequently Bought Together