ContactUsLinkComponent
PYRUVATE OXIDASE. From: Microorganism (EC 1.2.3.3) Lyophilized powder Activity: Not less than 1.5 units/mg-solid Unit Definition: One unit causes the formation of one micromole of hydrogen peroxide (half a micromole of quinoneimine dye) per minute 1 * 25 AU
Catalog # ICNA0215348625
Supplier:  MP Biomedicals
PYRUVATE OXIDASE. From: Microorganism (EC 1.2.3.3) Lyophilized powder Activity: Not less than 1.5 units/mg-solid Unit Definition: One unit causes the formation of one micromole of hydrogen peroxide (half a micromole of quinoneimine dye) per minute 1 * 25 AU
Catalog # ICNA0215348625
Supplier:  MP Biomedicals
Frequently Bought Together