ContactUsLinkComponent
Completely distinct from endoglycosidase F hydrolyzes the glycosylamine linkage of all asparagine-linked oligosacc harides. Substrates: Ribonuclease B, human transferrin, #-1 1 * 50 AU
Catalog # ICNA0215140650
Supplier:  MP Biomedicals
Completely distinct from endoglycosidase F hydrolyzes the glycosylamine linkage of all asparagine-linked oligosacc harides. Substrates: Ribonuclease B, human transferrin, #-1 1 * 50 AU
Catalog # ICNA0215140650
Supplier:  MP Biomedicals
Frequently Bought Together