ContactUsLinkComponent
MP ALUMINA B ACTIVITY SUPER I. Specific Surface Area: 200 m2 /g Particle Size: 63-200 µ m Weight: 0.8 g/ml pH: 10 1 * 50 kg
Catalog # ICNA0204577
Supplier:  MP Biomedicals
MP ALUMINA B ACTIVITY SUPER I. Specific Surface Area: 200 m2 /g Particle Size: 63-200 µ m Weight: 0.8 g/ml pH: 10 1 * 50 kg
Catalog # ICNA0204577
Supplier:  MP Biomedicals
Frequently Bought Together