ContactUsLinkComponent
MP ALUMINA B 18-32 µM, ACTIVE. Specific Surface Area: 200 m2 /gParticle Size: 18-32 µmWeight: 0.8 g/mlpH: 10.0 1 * 100 g
Catalog # ICNA0202065
Supplier:  MP Biomedicals
MP ALUMINA B 18-32 µM, ACTIVE. Specific Surface Area: 200 m2 /gParticle Size: 18-32 µmWeight: 0.8 g/mlpH: 10.0 1 * 100 g
Catalog # ICNA0202065
Supplier:  MP Biomedicals
Frequently Bought Together