ContactUsLinkComponent
For microbiological procedures. Vitamin Free This is a 10%, acid hydrolyzed, highly purified casein, processed to be free of the B complex vitamins. 1 * 500 mL
Catalog # ICNA0210476990
Supplier:  MP Biomedicals
For microbiological procedures. Vitamin Free This is a 10%, acid hydrolyzed, highly purified casein, processed to be free of the B complex vitamins. 1 * 500 mL
Catalog # ICNA0210476990
Supplier:  MP Biomedicals
Frequently Bought Together