ContactUsLinkComponent
Disodium Salt Tetrahydrate Crystalline Purity: 98-100% Substrate for CPK determination. Substantially free of creatine. 1 * 250 mg
  ICNA02100520.2
Disodium Salt Tetrahydrate Crystalline Purity: 98-100% Substrate for CPK determination. Substantially free of creatine. 1 * 250 mg
  ICNA02100520.2
Frequently Bought Together