ContactUsLinkComponent
NAPROXEN. Pharmacological agent that reportedly may show activity through ability to inhibit prostaglandin biosynthesis. 1 * 5 g
Catalog # ICNA0219024705
Supplier:  MP Biomedicals
NAPROXEN. Pharmacological agent that reportedly may show activity through ability to inhibit prostaglandin biosynthesis. 1 * 5 g
Catalog # ICNA0219024705
Supplier:  MP Biomedicals
Frequently Bought Together