MES MONOHYDRATE FREE ACID. Molecular Biology Reagent. Purity: >99% Zwitterionic bufferpKa = 6.15 at 25C 1 * 100 g
ICNA0219483580
MES MONOHYDRATE FREE ACID. Molecular Biology Reagent. Purity: >99% Zwitterionic bufferpKa = 6.15 at 25C 1 * 100 g
ICNA0219483580
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection