ContactUsLinkComponent
ALCOHOL OXIDASE. From Pichea pastoris E.C.1.1.3.13 Catalyzes the oxidation of an alcohol to an aldehyde and hydrogen peroxide. Solut ion in Sucrose. Activity: 10-20 units per mg protein. Unit Definition: One unit will oxidize 1.0 µmole of methanol to 1 * 1 KU
Catalog # ICNA0219015501
Supplier:  MP Biomedicals
ALCOHOL OXIDASE. From Pichea pastoris E.C.1.1.3.13 Catalyzes the oxidation of an alcohol to an aldehyde and hydrogen peroxide. Solut ion in Sucrose. Activity: 10-20 units per mg protein. Unit Definition: One unit will oxidize 1.0 µmole of methanol to 1 * 1 KU
Catalog # ICNA0219015501
Supplier:  MP Biomedicals
Frequently Bought Together