STAUROSPORINE. From Streptomyces sp. Purity: >98%.Inhibitor of phospholipid/Ca2+ dependent and cyclic nucleotide dependent protein kinases. A potent protein kinase C inhibitor and useful as a tool for studies on protein phosphorylation in the regulat 1 * 100 µG
Catalog # ICNA0219480580
Supplier: MP Biomedicals
Frequently Bought Together