ContactUsLinkComponent
(Phosphodiesterase 3,5´-cyclic-Nucleotide Activator).From Bovine Brain. Purity: >99% Lyophilized with 1.7 mM HEPES, pH7, 30 µM CaCl2 Activity: >13,000 units/mg protein Unit Definition: One unit will activate 3´ ,5´-cyclic-nucleotide phosphodiesterase 1 * 1 mg
Catalog # ICNA0219569101
Supplier:  MP Biomedicals
(Phosphodiesterase 3,5´-cyclic-Nucleotide Activator).From Bovine Brain. Purity: >99% Lyophilized with 1.7 mM HEPES, pH7, 30 µM CaCl2 Activity: >13,000 units/mg protein Unit Definition: One unit will activate 3´ ,5´-cyclic-nucleotide phosphodiesterase 1 * 1 mg
Catalog # ICNA0219569101
Supplier:  MP Biomedicals
Frequently Bought Together