ContactUsLinkComponent
n-BUTANOL. Molecular Biology Reagent.Purity: 99+% Useful for ethidium bromide removal from DNA purified by CsCl gradient ultracentrif ugation. It may also be used in the concentration of dilute nucleic acid solutions by repeated extractions. 1 * 1 L
Catalog # ICNA0219400191
Supplier:  MP Biomedicals
n-BUTANOL. Molecular Biology Reagent.Purity: 99+% Useful for ethidium bromide removal from DNA purified by CsCl gradient ultracentrif ugation. It may also be used in the concentration of dilute nucleic acid solutions by repeated extractions. 1 * 1 L
Catalog # ICNA0219400191
Supplier:  MP Biomedicals
Frequently Bought Together