ContactUsLinkComponent
5-BROMO-4-CHLORO-3-INDOLYL-beta-D-GLUCOPYRANOSIDE. Purity: 98%A -D -glucosidase substrate which renders an insoluble indigo-blue c hromophore (615 nm) upon enzymatic action. For histochemistry procedures, it acts as an indicator probe and detects 1 * 100 mg
Catalog # ICNA0219392380
Supplier:  MP Biomedicals
5-BROMO-4-CHLORO-3-INDOLYL-beta-D-GLUCOPYRANOSIDE. Purity: 98%A -D -glucosidase substrate which renders an insoluble indigo-blue c hromophore (615 nm) upon enzymatic action. For histochemistry procedures, it acts as an indicator probe and detects 1 * 100 mg
Catalog # ICNA0219392380
Supplier:  MP Biomedicals
Frequently Bought Together