ContactUsLinkComponent
Greater than 55 mg dexamethasone per gram with a balance consisting of 2-hydroxypropyl- -cyclodextrin. Typically used in cell culture applications. 1 * 50 mg
Catalog # ICNA02199350.6
Supplier:  MP Biomedicals
Greater than 55 mg dexamethasone per gram with a balance consisting of 2-hydroxypropyl- -cyclodextrin. Typically used in cell culture applications. 1 * 50 mg
Catalog # ICNA02199350.6
Supplier:  MP Biomedicals
Frequently Bought Together